Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MERCURY ACETATE 2.5% W/W IN GLACIAL ACETIC ACID BROWN GLASS FLASK 1 * 1 L
Catalog # PRLS89741.290
Supplier:
PROLABO CMP
Certificates
Expand image
MERCURY ACETATE 2.5% W/W IN GLACIAL ACETIC ACID BROWN GLASS FLASK 1 * 1 L
Catalog # PRLS89741.290
Supplier:
PROLABO CMP
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
pH electrodes, SenTix® PLUS
Supplier: WTW
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck