IRON CHLORIDE FOR G48 AND A923 METHODE C contains FeCl3 1 * 2,5 L
Catalog # PRLS89733.320
Supplier: PROLABO CMP
IRON CHLORIDE FOR G48 AND A923 METHODE C contains FeCl3 1 * 2,5 L
Catalog # PRLS89733.320
Supplier: PROLABO CMP
Frequently Bought Together
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Joint grease, K.W.S.
Supplier: POLLATH
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Hydrochloric acid 35%, TECHNICAL
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
MARBLE ETCHANT 1 * 1 L
Supplier: PROLABO CMP