CORROSION RCCM MC 1310 contains H2SO4 and CuSO4, 5 H2O 1 * 2,5 L
Catalog # PRLS89731.320
Supplier: PROLABO CMP
CORROSION RCCM MC 1310 contains H2SO4 and CuSO4, 5 H2O 1 * 2,5 L
Catalog # PRLS89731.320
Supplier: PROLABO CMP
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Copper ≥99.5%, turnings, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
pH electrodes, SenTix® PLUS
Supplier: WTW