MULTI ELEMENT STD 75 BLANK OIL S-21:30 1 * 200 g
Catalog # PROOCON150-021-027
Supplier: LGC Standards PROMOCHEM
MULTI ELEMENT STD 75 BLANK OIL S-21:30 1 * 200 g
Catalog # PROOCON150-021-027
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Macro Cuvettes
Supplier: VWR Collection
200g 1 * 200 g
Supplier: LGC Standards PROMOCHEM
Accessories for soxhlet extraction systems, 4 and 6 place
Supplier: GERHARDT