DNA containing double-strand breaks (DSB) created by the CRISPR/Cas9 system can be repaired by either the non-homologous end-joining (NHEJ) or the homology-directed repair (HDR) pathway. 1 * 100 µG
Catalog # BWRLBS8987
Supplier: Bioworld Technology
Frequently Bought Together