CD32 (also designated Fc gamma RII) is a low affinity receptor for the Fc fragment of aggregated IgG. CD32 is responsible for the cl earance of immunocomplexes by macrophages and also plays an important role in the regulation of antibody production b 1 * 100 µG
Catalog # BWRLBS8981
Supplier: Bioworld Technology
Frequently Bought Together