RBM29 (RNA binding motif protein 29), also known as ELMOD3 (ELMO domain-containing protein 3) or RBED1 (RNA-binding motif and ELMO d omain-containing protein 1), is a 381 amino acid protein that contains one ELMO domain and exists as six alternativel 1 * 100 µG
Catalog # BWRLBS8972
Supplier: Bioworld Technology
Frequently Bought Together