This gene is a member of the calcium/calmodulin-dependent protein kinase 1 family, a subfamily of the serine/threonine kinases. The encoded protein is a component of the calcium-regulated calmodulin-dependent protein kinase cascade. 1 * 100 µG
Catalog # BWRLBS8964
Supplier: Bioworld Technology
Frequently Bought Together