Order Entry
Netherlands
ContactUsLinkComponent
The transcriptional enhancer factor (TEF)/TEAD family includes TEF-1, TEF-3, TEF-4 and TEF-5. These proteins share a highly conserve d 68-amino acid TEA/ATTS DNA-binding domain, which binds to SV40 GT-IIC (GGAATG), SphI (AGTATG), SphII (AGCATG), and 1 * 100 µG
  BWRLBS61393
The transcriptional enhancer factor (TEF)/TEAD family includes TEF-1, TEF-3, TEF-4 and TEF-5. These proteins share a highly conserve d 68-amino acid TEA/ATTS DNA-binding domain, which binds to SV40 GT-IIC (GGAATG), SphI (AGTATG), SphII (AGCATG), and 1 * 100 µG
  BWRLBS61393
Frequently Bought Together