NDUFB1 (NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1), also known as CI-MNLL (complex I-MNLL), CI-SGDH or NADH-ubiqui none oxidoreductase MNLL subunit, is a 58 amino acid single-pass membrane protein that localizes to the matrix side o 1 * 100 µG
Catalog # BWRLBS61217
Supplier: Bioworld Technology
Frequently Bought Together