Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit.Together these sub units are composed of 4 RNA species and approximately 80 structurally distinct proteins. 1 * 100 µG
Catalog # BWRLBS61115
Supplier: Bioworld Technology
Frequently Bought Together