Order Entry
Netherlands
ContactUsLinkComponent
MP68, also known as PLPM or C14orf2 (chromosome 14 open reading frame 2), is a 58 amino acid mitochondrial protein that belongs to t he small mitochondrial proteolipid family. T 1 * 100 µG
Catalog # BWRLBS61111
MP68, also known as PLPM or C14orf2 (chromosome 14 open reading frame 2), is a 58 amino acid mitochondrial protein that belongs to t he small mitochondrial proteolipid family. T 1 * 100 µG
Catalog # BWRLBS61111
Frequently Bought Together