The human IL-3, IL-5 and GM-CSF receptors are each composed of both unique a subunits and a common β subunit. The a subunits are low affinity ligand binding proteins while the β subunits do not themselves bind ligand, but are required for high affin 1 * 100 µG
Catalog # BWRLBS60920
Supplier: Bioworld Technology
Frequently Bought Together