N-myristoyltransferase (NMT) catalyzes the reaction of N-terminal myristoylation of many signaling proteins. It transfers myristic a cid from myristoyl coenzyme A to the amino group of a protein's N-terminal glycine residue. 1 * 100 µG
Catalogus # BWRLBS8667
Supplier: Bioworld Technology
Frequently Bought Together