Order Entry
Netherlands
ContactUsLinkComponent
Kinetex® EVO C18 100 Å, LC Column, 30 x 2.1 mm 1 * 2,6 µm
Catalog # PHEN00A-4725-AN
Supplier:  PHENOMENEX
Kinetex® EVO C18 100 Å, LC Column, 30 x 2.1 mm 1 * 2,6 µm
Catalog # PHEN00A-4725-AN
Supplier:  PHENOMENEX
Frequently Bought Together