Order Entry
Netherlands
ContactUsLinkComponent
THESE OPTIMA 8X00 SERIES QUARTZ INJECTORS ARE USED FOR VOLATILE ORGANIC SOLVENT INTRODUCTION AND RECOMMENDED FOR A WIDE RANGE OF NON -HF APPLICATIONS. 1 * 1 items
  PERCN0782008
THESE OPTIMA 8X00 SERIES QUARTZ INJECTORS ARE USED FOR VOLATILE ORGANIC SOLVENT INTRODUCTION AND RECOMMENDED FOR A WIDE RANGE OF NON -HF APPLICATIONS. 1 * 1 items
  PERCN0782008
Frequently Bought Together