1.6MM ID SAMPLE LOOP, TEFLON, 500µL, CONNECTS TO PORTS, #1 AND #4 SC-FAST HIGH-FLOW (1/4-28 FITTINGS, 3.2MM OD X 1.6MM ID TUBING). 1 * 1 items
Catalog # PERCN0777832
Leverancier: PerkinElmer
1.6MM ID SAMPLE LOOP, TEFLON, 500µL, CONNECTS TO PORTS, #1 AND #4 SC-FAST HIGH-FLOW (1/4-28 FITTINGS, 3.2MM OD X 1.6MM ID TUBING). 1 * 1 items
Catalog # PERCN0777832
Leverancier: PerkinElmer
Frequently Bought Together
VWR®, Storage Boxes for Sample Vials/Bottles
Leverancier: VWR Collection
HPLC columns, LiChrospher®
Leverancier: Merck
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Leverancier: Avantor
VWR®, Screw Caps for Short Thread Vials ND9
Leverancier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND8
Leverancier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Leverancier: VWR Collection
Microliter™ 700 series syringes
Leverancier: HAMILTON BONADUZ
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Leverancier: VWR Collection
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 3 µm
Leverancier: Avantor
Avantor® ACE® Excel® C18-AR, HPLC/UHPLC Columns, Analytical, 3 µm
Leverancier: Avantor
Silica gel, Supelco®
Leverancier: Merck
Screw vial, fixed insert, amber 1 * 100 items
Leverancier: Agilent
ACQUITY UPLC BEH300 C4 1.7UM 2.1X150MM
Leverancier: WATERS
Caps for LC vials
Leverancier: Agilent
Exhaust filters, b.safe
Leverancier: Bohlender
Gas-clean filters
Leverancier: VARIAN
Canisters, S55, b.safe
Leverancier: Bohlender
Safety caps and safety waste cap accessories and replacement parts
Leverancier: S.C.A.T.
L-MARK® Replacement Needle for CTC Analytics PAL DLW (Dynamic Load and Wash)
Leverancier: Trajan Scientific and Medical