Order Entry
Netherlands
Orders LinkContactUsLinkComponent
VWR®, Erlenmeyer Flasks, with Screw Cap

VWR®, Erlenmeyer Flasks, with Screw Cap

100 ml, 64 mm, GL 25
Catalog # 214-0408
Supplier:   VWR Collection
VWR®, Erlenmeyer Flasks, with Screw Cap
Catalog # 214-0408
Supplier:  VWR Collection
VWR®, Erlenmeyer Flasks, with Screw Cap
VWR®, Erlenmeyer Flasks, with Screw Cap

100 ml, 64 mm, GL 25

Price
25,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate 3.3 glass, blue PP screw cap with PTFE liner.

  • Highly resistant to chemicals and thermal shock
  • Graduations and white labelling area
  • Delivered with blue PP screw cap

Ideal for storage or transportation of dangerous liquids. They are leakproof and can be autoclaved together with the screw cap at 140 °C.

Be aware that the screw cap should not be closed during the autoclaving process.

Specifications
  • Ext.Ø:
    64 mm
  • Thread:
    GL 25
  • Capacity:
    100 ml
  • Seq:
    02