PHASE MOBILE POUR HPLC PREPARATIVE 1 * 5 L
Catalog # PRLS89686.360
Supplier: PROLABO CMP
Frequently Bought Together
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Qualitative test paper phosphatesmo MI for phosphatase in milk
Supplier: MACHEREY-NAGEL
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test Kits, Cadmium, Spectroquant®, Supelco®
Supplier: Merck
Test kits, chloride, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
PerfeCTa® qPCR FastMix® II
Supplier: Quantabio
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach