Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
McFarland Standard Set ( 1 each of 0.5, 1.0, 2.0, 3.0, 4.0) 5 x 10ml 1 * 1 KIT
Catalog # PROBSD2350
Supplier:
PROLAB INSTRUMENTS
Certificates
Expand image
McFarland Standard Set ( 1 each of 0.5, 1.0, 2.0, 3.0, 4.0) 5 x 10ml 1 * 1 KIT
Catalog # PROBSD2350
Supplier:
PROLAB INSTRUMENTS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier: Merck
Reagents for chlorine determination using Lovibond® MD photometers
Supplier: Lovibond
pH buffers, ready to use
Supplier: WTW
VWR® Cryoboxes and Dividers
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals