Order Entry
Netherlands
ContactUsLinkComponent
Microscope cover glasses, 84x124mm, borosilicate glass D 263, thickness No. 1.5 (0.16-0.19 mm), 100pcs./cardboard box, MARIENFELD SUPERIOR 1 * 1.000 items
Catalog # MARI01029990933
Supplier:  MARIENFELD
Microscope cover glasses, 84x124mm, borosilicate glass D 263, thickness No. 1.5 (0.16-0.19 mm), 100pcs./cardboard box, MARIENFELD SUPERIOR 1 * 1.000 items
Catalog # MARI01029990933
Supplier:  MARIENFELD
Frequently Bought Together