SEL ADOUCISSEUR 1 * 25 kg
Catalogus # PRLS89634.460
Supplier: PROLABO CMP
Frequently Bought Together
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Test kits, chromate, Spectroquant®, Supelco®
Supplier: Merck
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips for nickel, MQuant®, Supelco®
Supplier: Merck
Haemalum (Mayer's), Sigma-Aldrich®
Supplier: Merck
Lugol's iodine solution, Sigma-Aldrich®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Chinese ink, green, Q Path® stain for histology
Supplier: VWR Chemicals
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
Orthophosphoric acid 10%, TECHNICAL
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Supelco® Spectroquant® Test Kits, Sulphate
Supplier: Merck
Test kits, photometric, Sulphate, LCK cuvette tests
Supplier: Hach