Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Crucible, tall form

Crucible, tall form

Tall form, 200 ml, 73×81 mm
Catalogus # TSLA75106
Crucible, tall form
Catalogus # TSLA75106
Crucible, tall form

Tall form, 200 ml, 73×81 mm

Price
130,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Quartz, transparent, glazed.

Specificaties
  • Ø×H:
    73×81 mm
  • Type:
    Tall form
  • Capacity:
    200 ml
  • Seq:
    07
Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.