Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Crucible, tall form

Crucible, tall form

Tall form, 24 ml, 35×44 mm
Catalogus # TSLA75092
Crucible, tall form
Catalogus # TSLA75092
Crucible, tall form

Tall form, 24 ml, 35×44 mm

Price
23,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Quartz, transparent, glazed.

Specificaties
  • Ø×H:
    35×44 mm
  • Type:
    Tall form
  • Capacity:
    24 ml
  • Seq:
    02
Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.