Order Entry
Netherlands
ContactUsLinkComponent
Electrode Filling Solution, KCl 3M + AgCl (Crisolyt A), 125 ml 1 * 125 mL
Catalog # HACHLZW9511.99
Leverancier:  Hach
Electrode Filling Solution, KCl 3M + AgCl (Crisolyt A), 125 ml 1 * 125 mL
Catalog # HACHLZW9511.99
Leverancier:  Hach
Gerelateerde informatie
Frequently Bought Together