Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Conductivity Standard Solution, KCl 1.0M, 111.8 mS/cm, 250 ml with Certificate of Analysis (InterNet download) 1 * 250 mL
Catalog # HACHLZW9740.99
Leverancier:
Hach
Expand image
Conductivity Standard Solution, KCl 1.0M, 111.8 mS/cm, 250 ml with Certificate of Analysis (InterNet download) 1 * 250 mL
Catalog # HACHLZW9740.99
Leverancier:
Hach
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Leverancier: Merck
CONDUCTIVITY STANDARD 1.3µS/CM 1 * 250 mL
Leverancier: HAMILTON BONADUZ
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Leverancier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Leverancier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Leverancier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Leverancier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Leverancier: VWR Chemicals
pH buffers, ready to use
Leverancier: WTW
VWR®, Conductivity standard solutions
Leverancier: VWR Chemicals
pH buffers, DuraCal
Leverancier: HAMILTON BONADUZ