Oasis HLB 20cc Vac RC Cartridge, 30mg Sorbent per Cartridge, 30µm Particle Size 1 * 50 items
WATE186000382
: WATERS
Oasis HLB 20cc Vac RC Cartridge, 30mg Sorbent per Cartridge, 30µm Particle Size 1 * 50 items
WATE186000382
: WATERS
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals