Order Entry
Netherlands
ContactUsLinkComponent
CF680R picolyl azide reacts with alkyne to form 1,2,3-triazole by 1,3-dipolar Huisgen cycloaddition through the use of a much lower copper (I) concentration without sacrificing reaction efficiency. 1 * 0,5 mg
Catalog # 96007.
Supplier:  Biotium
CF680R picolyl azide reacts with alkyne to form 1,2,3-triazole by 1,3-dipolar Huisgen cycloaddition through the use of a much lower copper (I) concentration without sacrificing reaction efficiency. 1 * 0,5 mg
Catalog # 96007.
Supplier:  Biotium
Frequently Bought Together