BIOTIN - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO RAT IgG (WHOLE MOLECULE). Anti-Rat Host: goat Form: affinity purified liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. Biotinylat 1 * 2 mL
Catalog # ICNA0855790
Supplier: MP Biomedicals
Frequently Bought Together