Order Entry
Netherlands
ContactUsLinkComponent
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO RAT IgG (WHOLE MOLECULE). Anti-Rat Host: goat Form: lyophilized (IgG) Applications: Imm unostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855770
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO RAT IgG (WHOLE MOLECULE). Anti-Rat Host: goat Form: lyophilized (IgG) Applications: Imm unostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855770
Supplier:  MP Biomedicals
Frequently Bought Together