Order Entry
Netherlands
ContactUsLinkComponent
PEROXIDASE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO RABBIT IgG F(ab')2. Anti-Rabbit Host: goat Form: affinity purified liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conj 1 * 2 mL
Catalog # ICNA0855691
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO RABBIT IgG F(ab')2. Anti-Rabbit Host: goat Form: affinity purified liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conj 1 * 2 mL
Catalog # ICNA0855691
Supplier:  MP Biomedicals
Frequently Bought Together