Order Entry
Netherlands
ContactUsLinkComponent
FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO RABBIT IMMUNOGLOBULINS (IgG, IgA, IgM). (Anti-IgG, IgA, and IgM) Anti-Rabbit Host: goa t Form: lyophilized (IgG) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded 1 * 2 mL
Catalog # ICNA0855652
Supplier:  MP Biomedicals
FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO RABBIT IMMUNOGLOBULINS (IgG, IgA, IgM). (Anti-IgG, IgA, and IgM) Anti-Rabbit Host: goa t Form: lyophilized (IgG) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded 1 * 2 mL
Catalog # ICNA0855652
Supplier:  MP Biomedicals
Frequently Bought Together