Order Entry
Netherlands
ContactUsLinkComponent
PEROXIDASE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO MOUSE IgG Fc. Anti-Mouse Host: goat Form: affinity purified liquid Applic ations: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855566
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO MOUSE IgG Fc. Anti-Mouse Host: goat Form: affinity purified liquid Applic ations: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855566
Supplier:  MP Biomedicals
Frequently Bought Together