Order Entry
Netherlands
ContactUsLinkComponent
PEROXIDASE - CONJUGATED SHEEP IgG FRACTION TO MOUSE IgG (NO CROSS TO HUMAN). Anti-Mouse Host: sheep Form: lyophilized (IgG) Applicat ions: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. This antibody is 1 * 2 mL
Catalog # ICNA0855558
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED SHEEP IgG FRACTION TO MOUSE IgG (NO CROSS TO HUMAN). Anti-Mouse Host: sheep Form: lyophilized (IgG) Applicat ions: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. This antibody is 1 * 2 mL
Catalog # ICNA0855558
Supplier:  MP Biomedicals
Frequently Bought Together