Order Entry
Netherlands
ContactUsLinkComponent
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO MOUSE IMMUNOGLOBULINS (IgG, IgA, IgM). (Anti-IgG, IgA, and IgM) Anti-Mouse Host: goat F orm: lyophilized (IgG) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tis 1 * 2 mL
Catalog # ICNA0855556
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO MOUSE IMMUNOGLOBULINS (IgG, IgA, IgM). (Anti-IgG, IgA, and IgM) Anti-Mouse Host: goat F orm: lyophilized (IgG) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tis 1 * 2 mL
Catalog # ICNA0855556
Supplier:  MP Biomedicals
Frequently Bought Together