Order Entry
Netherlands
ContactUsLinkComponent
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: goat Form: lyophilized IgG Applications: I mmunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855550
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: goat Form: lyophilized IgG Applications: I mmunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855550
Supplier:  MP Biomedicals
Frequently Bought Together