Order Entry
Netherlands
ContactUsLinkComponent
FITC CONJUGATED GOAT AFFINITY PURIFIED IgG TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: goat Form: affinity purified liquid Appli cations: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugate 1 * 2 mg
Catalog # ICNA0855514
Supplier:  MP Biomedicals
FITC CONJUGATED GOAT AFFINITY PURIFIED IgG TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: goat Form: affinity purified liquid Appli cations: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugate 1 * 2 mg
Catalog # ICNA0855514
Supplier:  MP Biomedicals
Frequently Bought Together