Order Entry
Netherlands
ContactUsLinkComponent
FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO MOUSE IgG F(ab')2. Anti-Mouse Host: goat Form: lyophilized (IgG) Applications: Immunos taining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855496
Supplier:  MP Biomedicals
FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO MOUSE IgG F(ab')2. Anti-Mouse Host: goat Form: lyophilized (IgG) Applications: Immunos taining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855496
Supplier:  MP Biomedicals
Frequently Bought Together