SHEEP AFFINITY PURIFIED ANTIBODY TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: sheep Form: affinity purified liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mg
Catalog # ICNA0855481
Supplier: MP Biomedicals
Frequently Bought Together