Order Entry
Netherlands
ContactUsLinkComponent
GOAT AFFINITY PURIFIED ANTIBODY TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: goat Form: affinity purified liquid Applications: Im munostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mg
Catalog # ICNA0855479
Supplier:  MP Biomedicals
GOAT AFFINITY PURIFIED ANTIBODY TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: goat Form: affinity purified liquid Applications: Im munostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mg
Catalog # ICNA0855479
Supplier:  MP Biomedicals
Frequently Bought Together