Order Entry
Netherlands
ContactUsLinkComponent
PEROXIDASE - CONJUGATED RABBIT AFFINITY PURIFIED ANTIBODY TO GOAT IgG (WHOLE MOLECULE). Anti-Goat Host: rabbit Form: affinity purifi ed liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855363
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED RABBIT AFFINITY PURIFIED ANTIBODY TO GOAT IgG (WHOLE MOLECULE). Anti-Goat Host: rabbit Form: affinity purifi ed liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855363
Supplier:  MP Biomedicals
Frequently Bought Together