Order Entry
Netherlands
ContactUsLinkComponent
FLUORESCEIN-CONJUGATED GOAT IgG FRACTION TO DOG IgM (µ CHAIN). Anti-DogHost: goatForm: lyophilized (IgG)Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC Conjugated 1 * 2 mL
Catalog # ICNA0855328
Supplier:  MP Biomedicals
FLUORESCEIN-CONJUGATED GOAT IgG FRACTION TO DOG IgM (µ CHAIN). Anti-DogHost: goatForm: lyophilized (IgG)Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC Conjugated 1 * 2 mL
Catalog # ICNA0855328
Supplier:  MP Biomedicals
Frequently Bought Together