FLUORESCEIN - CONJUGATED RABBIT IgG FRACTION TO DOG IgG (WHOLE MOLECULE). Anti-Dog Host: rabbit Form: lyophilized (IgG) Applications : Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855326
Supplier: MP Biomedicals
Frequently Bought Together