Order Entry
Netherlands
ContactUsLinkComponent
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO HUMAN IgG Fc. Anti-Human Host: goat Form: lyophilized (IgG) Applications: Immunostainin g of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855226
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO HUMAN IgG Fc. Anti-Human Host: goat Form: lyophilized (IgG) Applications: Immunostainin g of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855226
Supplier:  MP Biomedicals
Frequently Bought Together