Order Entry
Netherlands
ContactUsLinkComponent
PEROXIDASE - CONJUGATED RABBIT IgG FRACTION TO HUMAN IgG (WHOLE MOLECULE). Anti-Human Host: rabbit Form: lyophilized (IgG) Applicati ons: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855221
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED RABBIT IgG FRACTION TO HUMAN IgG (WHOLE MOLECULE). Anti-Human Host: rabbit Form: lyophilized (IgG) Applicati ons: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855221
Supplier:  MP Biomedicals
Frequently Bought Together