FITC CONJUGATED GOAT AFFINITY PURIFIED IgG TO HUMAN IgG (WHOLE MOLECULE). Anti-Human Host: goat Form: affinity purified liquid Appli cations: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugate 1 * 2 mg
Catalog # ICNA0855197
Supplier: MP Biomedicals
Frequently Bought Together