Order Entry
Netherlands
ContactUsLinkComponent
FLUORESCEIN-CONJUGATED GOAT F(ab')2 FRAGMENT TO HUMAN IgA (alpha CHAIN). Anti-HumanHost: goatForm: lyophilized (F(ab)2 )Application s: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC Conjugated 1 * 2 mL
Catalog # ICNA0855178
Supplier:  MP Biomedicals
FLUORESCEIN-CONJUGATED GOAT F(ab')2 FRAGMENT TO HUMAN IgA (alpha CHAIN). Anti-HumanHost: goatForm: lyophilized (F(ab)2 )Application s: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC Conjugated 1 * 2 mL
Catalog # ICNA0855178
Supplier:  MP Biomedicals
Frequently Bought Together