Order Entry
Netherlands
ContactUsLinkComponent
FLUORESCEIN-CONJUGATED GOAT IgG FRACTION TO HUMAN LAMBDA LIGHT CHAIN (BOUND). Anti-HumanHost: goatForm: lyophilized (IgG)Application s: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC ConjugatedGoat P 1 * 2 mL
Catalog # ICNA0855160
Supplier:  MP Biomedicals
FLUORESCEIN-CONJUGATED GOAT IgG FRACTION TO HUMAN LAMBDA LIGHT CHAIN (BOUND). Anti-HumanHost: goatForm: lyophilized (IgG)Application s: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC ConjugatedGoat P 1 * 2 mL
Catalog # ICNA0855160
Supplier:  MP Biomedicals
Frequently Bought Together