Order Entry
Netherlands
ContactUsLinkComponent
GOAT F(ab')2 FRAGMENT TO HUMAN IgM (5Fc µ). Anti-Human Host: goat Form: lyophilized F(ab´)2 Applications: Immunostaining of acetone- fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855055
Supplier:  MP Biomedicals
GOAT F(ab')2 FRAGMENT TO HUMAN IgM (5Fc µ). Anti-Human Host: goat Form: lyophilized F(ab´)2 Applications: Immunostaining of acetone- fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855055
Supplier:  MP Biomedicals
Frequently Bought Together