Order Entry
Netherlands
ContactUsLinkComponent
GOAT IgG FRACTION TO HUMAN IgM (5Fc µ). Anti-Human Host: goat Form: lyophilized (IgG) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 5 mL
Catalog # ICNA0855017
Supplier:  MP Biomedicals
GOAT IgG FRACTION TO HUMAN IgM (5Fc µ). Anti-Human Host: goat Form: lyophilized (IgG) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 5 mL
Catalog # ICNA0855017
Supplier:  MP Biomedicals
Frequently Bought Together